Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144134 Supplier Invitrogen™ Supplier No. PA5144134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C2orf83 Polyclonal Antibody for IHC (P), ELISA

C2orf83 (Chromosome 2 Open Reading Frame 83) is a Protein Coding gene. An important paralog of this gene isSLC19A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf83
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene C2orf83
Gene Accession No. Q53S99
Gene Alias C2orf83; chromosome 2 open reading frame 83; folate transporter-like protein C2orf83
Gene Symbols C2orf83
Host Species Rabbit
Immunogen Recombinant Human Folate transporter-like protein C2orf83 protein (29-150AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56918
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.