Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf89 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989373
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9373 Supplier Bioss Supplier No. BS9820R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Metalloprotease that acts as a negative regulator of the Wnt signaling pathway by mediating the cleavage of the 8 N- terminal residues of a subset of Wnt proteins. Following cleavage, Wnt proteins become oxidized and form large disulfide-bond oligomers, leading to their inactivation. Able to cleave WNT3A, WNT5, but not WNT11. Required for head formation.

Specifications

Antigen C2orf89
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRABD2A
Gene Accession No. Q86V40
Gene Alias C2orf89; Metalloprotease TIKI1; TIKI1; TraB domain containing 2A; TRAB domain-containing protein 2A; TRABD2A; UPF0632 protein C2orf89
Gene Symbols TRABD2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C2orf89.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129293
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.