Learn More
C3/C3b/C3c Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 213371AP150UL

Description
This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.Specifications
C3/C3b/C3c | |
Polyclonal | |
Unconjugated | |
C3 | |
ARMD9, ASP, C3, C3b, C3c, Complement C3, complement component 3, CPAMD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24232, 718 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01024, P01026 | |
C3 | |
C3/C3b/C3c Fusion Protein Ag15537 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.