Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3 Goat anti-Guinea pig, HRP, Polyclonal , Abnova™

Catalog No. 89933693 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-693 Supplier Abnova Corporation Supplier No. PAB29724

Goat polyclonal antibody raised against guinea pig C3.

Specifications

Antigen C3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Goat polyclonal antibody raised against guinea pig C3.
Dilution ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM HEPES (Stabilizer, 0.002% Thimerosal)
Gene Symbols C3
Host Species Goat
Immunogen Native purified guinea pig C3.
Purification Method Antigen Affinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135462
Target Species Guinea Pig
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.