Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3, Rabbit anti-Human, Clone: E28-P, Abnova™

Catalog No. 89999154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-154 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-154 Supplier Abnova Corporation Supplier No. MAB16396
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen C3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E28-P
Conjugate Unconjugated
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Immunohistochemistry (Frozen sections) The optimal working dilution should be determined by the end user.
Formulation In 20 mM Tris-HCl, pH 8.0 (20 mg/mL BSA, 0.05% sodium azide).
Gene complement component 3
Gene Alias ARMD9/ASP/CPAMD1
Gene Symbols C3
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human Complement C3d.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.