Learn More
Invitrogen™ C3a Receptor Monoclonal Antibody (74), FITC

Description
MA5-17475 targets C3a Receptor in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17475 immunogen is rBL-2H3 transfectants expressing rat C3aR.
Specifications
Specifications
| Antigen | C3a Receptor |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 74 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | C3ar1 |
| Gene Accession No. | O55197 |
| Gene Alias | anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.