Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3a Receptor Monoclonal Antibody (74)
GREENER_CHOICE

Catalog No. PIMA517473
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517473 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517473 Supplier Invitrogen™ Supplier No. MA517473
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA5-17473 targets C3a Receptor in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17473 immunogen is rBL-2H3 transfectants expressing rat C3aR.

C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a Receptor
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 74
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene C3ar1
Gene Accession No. O55197
Gene Alias anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09
Gene Symbols C3ar1
Host Species Mouse
Immunogen RBL-2H3 transfectants expressing rat C3aR.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84007
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.