Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3a Receptor Mouse anti-Human, RY586, Clone: hC3aRZ8, BD OptiBuild™

Catalog No. BDB753121 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB753121 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB753121 Supplier BD Biosciences Supplier No. 753121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C3a Receptor Monoclonal antibody specifically detects C3a Receptor in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone hC3aRZ8
Concentration 0.2 mg/mL
Conjugate RY586
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias C3AR; C3a-R; C3AR1; C3a anaphylatoxin chemotactic receptor; AZ3B
Host Species Mouse
Immunogen Human C3aR Transfected Cell Line
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.