Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C3aR Rat anti-Mouse, BUV 805, Clone: 14D4, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 753368
This item is not returnable.
View return policy
Description
C3aR Monoclonal antibody specifically detects C3aR in Mouse samples. It is validated for Flow Cytometry
Specifications
C3aR | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Flow Cytometry | |
14D4 | |
Brilliant Ultraviolet 805 | |
C3AR; C3a-R; C3ar1; anaphylatoxin C3a receptor | |
Mouse C3aR Transfected Cell Line | |
RUO | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction