Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3IP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3516 Supplier Bioss Supplier No. BS15164R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.

Specifications

Antigen C3IP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Klhl12
Gene Accession No. Q53G59
Gene Alias BOS_15933; C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like 12 (Drosophila); kelch-like family member 12; Kelch-like protein 12; kelch-like protein C3IP1; klhl12; si:dkeyp-53e12.5; wu:fi60g03; zgc:92570
Gene Symbols Klhl12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3IP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59349
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.