Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C3orf10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543671
Description
Peptide sequence: YIEIITSSIK KIADFLNSFD MSCRSRLATL NEKLTALERR IEYIEARVTK Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 100%.
C3orf10 is involved in regulation of actin and microtubule organization. It is a part of a WAVE complex that activates the Arp2/3 complex.
Specifications
C3orf10 | |
Polyclonal | |
Unconjugated | |
BRK1 | |
6720456B07Rik; AW011779; BRICK1 subunit of SCAR/WAVE actin nucleating complex; BRICK1, SCAR/WAVE actin nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit, homolog; BRK1; C22H3orf10; C3orf10; haematopoietic stem/progenitor cell protein 300; hHBrk1; HSPC300; MDS027; Probable protein BRICK1; protein BRICK1; protein BRICK1-like protein; zgc:86903 | |
Rabbit | |
Protein A | |
RUO | |
55845 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WUW1 | |
BRK1 | |
Synthetic peptide directed towards the middle region of human C3orf10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction