Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566323 Supplier Invitrogen™ Supplier No. PA566323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGQEDPVQRE IHQDWANREY IEIITSSIKK IADFLNSFDM SCRSRLATLN EKLTALERRI EYIEARVT Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

C3orf10 is involved in regulation of actin and microtubule organization. It is a part of a WAVE complex that activates the Arp2/3 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRK1
Gene Accession No. Q8WUW1
Gene Alias 6720456B07Rik; AW011779; BRICK1 subunit of SCAR/WAVE actin nucleating complex; BRICK1, SCAR/WAVE actin nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit, homolog; BRK1; C22H3orf10; C3orf10; haematopoietic stem/progenitor cell protein 300; hHBrk1; HSPC300; MDS027; Probable protein BRICK1; protein BRICK1; protein BRICK1-like protein; zgc:86903
Gene Symbols BRK1
Host Species Rabbit
Immunogen Recombinant Human C3orf10. Recombinant protein control fragment (Product #RP-107004).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.