Learn More
C3orf15 Mouse anti-Human, Clone: OTI7F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802487

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C3orf15 (MAATS1, MYCBP Associated And Testis Expressed 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include adenosylmethionine decarboxylase activity.Specifications
C3orf15 | |
Monoclonal | |
Unconjugated | |
MAATS1 | |
AAT1, AAT1alpha, C3orf15, CFAP91, CaM-IP2, SPATA26 | |
Mouse | |
Affinity Chromatography | |
RUO | |
89876 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q7Z4T9 | |
MAATS1 | |
Human recombit protein fragment corresponding to amino acids 1-313 of human C3orf15 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.