Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567059 Supplier Invitrogen™ Supplier No. PA567059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTFPSDITEFL GQPYFEAFKK KMPIAFAAKG INKLLNKLFL INEQSPRASE ETLLGISKKA KQMKINVQNN VDLG Highest antigen sequence identity to the following orthologs - mouse 59%, rat 52%.

May play a role in cortex development as part of the Notch signaling pathway. Downstream of Notch may repress the expression of proneural genes and inhibit neuronal differentiation thereby maintaining neural progenitors. May also play a role in preimplentation embryo development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf17
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEPRO
Gene Accession No. Q6NW34
Gene Alias BC027231; C33H3orf17; C3orf17; cDNA sequence BC027231; hypothetical protein MGC27931; NEPRO; NET17; Neural progenitor protein; Nucleolus and neural progenitor protein; protein nepro homolog; RGD1311458; similar to cDNA sequence BC027231; hypothetical protein MGC27931; uncharacterized protein C3orf17; uncharacterized protein C3orf17 homolog
Gene Symbols NEPRO
Host Species Rabbit
Immunogen Recombinant Human C3orf17. Recombinant protein control fragment (Product #RP-105481).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25871
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.