Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C3orf17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542636
Description
Peptide sequence: KQLLNSGVSM PVIQTKEKMI HENLRGIHEN ETDSWTVMQI NKNSTSGTIK Sequence homology: Dog: 79%; Human: 100%.
May play a role in cortex development as part of the Notch signaling pathway. Downstream of Notch may repress the expression of proneural genes and inhibit neuronal differentiation thereby maintaining neural progenitors. May also play a role in preimplentation embryo development.
Specifications
C3orf17 | |
Polyclonal | |
Unconjugated | |
NEPRO | |
BC027231; C33H3orf17; C3orf17; cDNA sequence BC027231; hypothetical protein MGC27931; NEPRO; NET17; Neural progenitor protein; Nucleolus and neural progenitor protein; protein nepro homolog; RGD1311458; similar to cDNA sequence BC027231; hypothetical protein MGC27931; uncharacterized protein C3orf17; uncharacterized protein C3orf17 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25871 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6NW34 | |
NEPRO | |
Synthetic peptide directed towards the middle region of human C3orf17. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction