Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989377
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9377 Supplier Bioss Supplier No. BS9824R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in cortex development as part of the Notch signaling pathway. Downstream of Notch may repress the expression of proneural genes and inhibit neuronal differentiation thereby maintaining neural progenitors. May also play a role in preimplentation embryo development.

Specifications

Antigen C3orf17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NEPRO
Gene Accession No. Q6NW34
Gene Alias C33H3orf17; C3orf17; chromosome 33 open reading frame, human C3orf17; NEPRO; NET17; Nucleolus and neural progenitor protein; protein nepro homolog; uncharacterized protein C3orf17
Gene Symbols NEPRO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf17.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25871
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.