Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C3orf18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113528
Description
C3orf18 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications
C3orf18 | |
Polyclonal | |
Unconjugated | |
C3orf18 | |
C3orf18; chromosome 3 open reading frame 18; G20; Protein G20; Uncharacterized protein C3orf18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51161 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9UK00 | |
C3orf18 | |
Recombinant Human Uncharacterized protein C3orf18 protein (83-162AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction