Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf20 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA558846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558846 Supplier Thermo Scientific Supplier No. PA558846

Rabbit Polyclonal Antibody

Immunogen sequence: CLSFSLSAGK EAKKKIGKSR TTEDVSMPPL HRGVGTPANS LEFSDPCPEA REKLQELCRH IEAERTTWKG RNISYPMILR NYKAKMPSHL MLARKGDSQT Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 59%.

The function of C3orf20 has not been widely studied, and is yet to be fully elucidated. Two isoforms were produced by alternative splicing with predicted MW of 88 and 101 kDa according to UniProt.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf20
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C3orf20
Gene Accession No. Q8ND61
Gene Alias C3orf20; chromosome 3 open reading frame 20; uncharacterized protein C3orf20
Gene Symbols C3orf20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C3orf20
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.