Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf21/XXYLT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989379
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9379 Supplier Bioss Supplier No. BS9826R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XXYLT1 gene ontology annotations related to this gene include transferase activity, transferring glycosyl groups.

Specifications

Antigen C3orf21/XXYLT1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene XXYLT1
Gene Accession No. Q8NBI6
Gene Alias AI480653; BOS_930; C1H3orf21; C3orf21; im:6912235; PSEC0251; RGD1308154; si:dkey-25g11.2; UDP-xylose:alpha-xyloside alpha-1,3-xylosyltransferase; uncharacterized protein C3orf21 homolog; Xxylt1; Xyloside xylosyltransferase 1
Gene Symbols XXYLT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152002
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.