Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C3orf22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569901
Description
This target displays homology in the following species: Cow: 83%; Human: 100%.
C3orf22 is a protein coding gene.
Specifications
C3orf22 | |
Polyclonal | |
Unconjugated | |
C3orf22 | |
C3orf22; chromosome 3 open reading frame 22; uncharacterized protein C3orf22 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
152065 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N5N4 | |
C3orf22 | |
synthetic peptide directed towards the middle region of human C3orf22 (aa 50-99). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction