Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf25 (EFCAB12) Monoclonal Antibody (OTI1A3), OriGene
SDP

Catalog No. 502543877
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3877 Supplier OriGene Supplier No. CF811950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C3orf25 (EFCAB12) Monoclonal antibody specifically detects C3orf25 (EFCAB12) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen C3orf25 (EFCAB12)
Applications Western Blot
Classification Monoclonal
Clone OTI1A3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene RFC3
Gene Accession No. P40938, Q8R323
Gene Alias C3orf25; EFCAB12; EF-hand calcium binding domain 12; EF-hand calcium-binding domain-containing protein 12; EF-hand domain-containing protein C3orf25
Host Species Mouse
Immunogen Full length human recombinant protein of human RFC3 (NP_002906) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288414, 5983, 69263
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.