Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560138 Supplier Invitrogen™ Supplier No. PA560138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEWWENQPTG AGSSPEASDG EGEGDTEVMQ QETVPVPVPS EKTKQPKECF LIQPKERKEN TTKTRKRRKK KITDVLAKSE PKP Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 54%.

CMSS1 (Cms1 Ribosomal Small Subunit Homolog) is a Protein Coding gene. Diseases associated with CMSS1 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf26
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMSS1
Gene Accession No. Q9BQ75
Gene Alias 1110001A06Rik; 2610528E23Rik; 4930572F24Rik; AV099812; AV122629; C3orf26; cms small ribosomal subunit 1; cms1 ribosomal small subunit homolog; cms1 ribosomal small subunit homolog (yeast); Cmss1; MGC4308; Protein CMSS1; RGD1309437
Gene Symbols CMSS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C3orf26. Recombinant protein control fragment (Product #RP-100262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.