Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3Orf34 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502206306
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6306 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6306 Supplier Fabgennix Inc Supplier No. C3ORF34101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C3Orf34 Polyclonal antibody specifically detects C3Orf34 in Human samples. It is validated for Dot Blot, ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen C3Orf34
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CEP19
Gene Accession No. Q96LK0
Gene Alias 1500031L02Rik; AI428934; AL022620; C3orf34; centrosomal protein 19; centrosomal protein 19kDa; centrosomal protein of 19 kDa; Cep19; HSD5; MOSPGF; RGD621352; testis specific protein
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 120-170 on human Chromosome open reading frame 34 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84984
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.