Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C3orf36 Antibody (OTI8G4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272218B
Description
C3orf36 Monoclonal specifically detects C3orf36 in Human samples. It is validated for Western Blot.Specifications
C3orf36 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
C3orf36 | |
Full length recombinant protein of human C3orf36 (NP_079317) produced in E.coli. | |
0.1 mL | |
80111.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI8G4 | |
Western Blot | |
C3orf36 chromosome 3 open reading frame 36 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction