Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C3orf36 Antibody (OTI8G4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272218PE
Description
C3orf36 Monoclonal antibody specifically detects C3orf36 in Human samples. It is validated for Western BlotSpecifications
C3orf36 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
80111 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
OTI8G4 | |
Western Blot | |
C3orf36 chromosome 3 open reading frame 36 | |
Full length human recombinant protein of human C3orf36(NP_079317) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction