Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf36 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983528
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3528 Supplier Bioss Supplier No. BS15176R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen C3orf36
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C3orf36
Gene Accession No. Q3SXR2
Gene Alias C3orf36; chromosome 3 open reading frame 36; uncharacterized protein C3orf36
Gene Symbols C3orf36
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf36.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80111
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.