Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf37 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989381
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9381 Supplier Bioss Supplier No. BS9829R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically binds 5-hydroxymethylcytosine (5hmC)-containing DNA in stem cells, suggesting that it acts as a specific reader of 5hmC in stem cells. May act as a peptidase; experimental evidences are however required to confirm this prediction.

Specifications

Antigen C3orf37
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HMCES
Gene Accession No. Q96FZ2
Gene Alias 5-hydroxymethylcytosine (hmC) binding, ES cell specific; 5-hydroxymethylcytosine (hmC) binding, ES cell-specific; 8430410A17Rik; Abasic site processing protein HMCES; BOS_21232; C20H3orf37; C22H3orf37; C3orf37; C85376; chromosome 22 open reading frame, human C3orf37; DC12; Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; ES cell-specific 5hmC-binding protein; Hmces; Peptidase HMCES; Putative endonuclease HMCES; putative peptidase SRAPD1; SRAP domain-containing protein 1; Srap1; Srapd1; UPF0361 protein C3orf37; UPF0361 protein C3orf37 homolog
Gene Symbols HMCES
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf37.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56941
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.