Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C3orf38 Rabbit anti-Human, Clone: S08-3H6, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
| Antigen | C3orf38 |
|---|---|
| Clone | S08-3H6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
C3orf38 Monoclonal antibody specifically detects C3orf38 in Human samples. It is validated for Western BlotSpecifications
| C3orf38 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 285237 | |
| IgG | |
| Affinity-purified |
| S08-3H6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| chromosome 3 open reading frame 38, FLJ54270, hypothetical protein LOC285237, MGC26717 | |
| Recombinant protein of human C3orf38 (Uniprot#: Q5JPI3) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title