Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf39 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9382 Supplier Bioss Supplier No. BS9830R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen C3orf39
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POMGNT2
Gene Accession No. Q8NAT1
Gene Alias Ago61; BOS_20684; C22H3orf39; C23H3orf39; C3orf39; C85492; Eogtl; Extracellular O-linked N-acetylglucosamine transferase-like; glycosyltransferase; glycosyltransferase ago61; glycosyltransferase-like; glycosyltransferase-like domain containing 2; Glycosyltransferase-like domain-containing protein 2; Gtdc2; LOC100341347; MDDGA8; POMGNT2; protein O-linked mannose beta 1,4-N-acetylglucosaminyltransferase 2; protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-); Protein O-linked-mannose beta-1,4-N-acetylglucosaminyltransferase 2; RGD1304827
Gene Symbols POMGNT2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf39.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84892
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.