Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf58 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983531
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3531 Supplier Bioss Supplier No. BS15179R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIPK2A (CC058, 1190002N15Rik) may play a role in cardiomyocyte proliferation through paracrine signaling and activation of the PPI3K-AKT-CDK7 signaling cascade

Specifications

Antigen C3orf58
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DIPK2A
Gene Accession No. Q8NDZ4
Gene Alias 1190002N15Rik; Ab2-095; BOS_1273; C1H3orf58; C23H3orf58; C3orf58; chromosome 1 open reading frame, human C3orf58; chromosome 23 open reading frame, human C3orf58; chromosome 3 open reading frame 58; deleted in autism 1; Deleted in autism protein 1; Deleted in autism protein 1 homolog; DIA1; DIPK2A; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; hypoxia and Akt induced stem cell factor; Hypoxia and AKT-induced stem cell factor; RGD1309079; RIKEN cDNA 1190002N15 gene; similar to Ab2-095; UPF0672 protein C3orf58; UPF0672 protein C3orf58 homolog
Gene Symbols DIPK2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C3orf58.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 205428
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.