Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569945 Supplier Invitrogen™ Supplier No. PA569945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

MB21D2 gene ontology annotations related to this gene include cadherin binding; protein-containing complex binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf59
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MB21D2
Gene Accession No. Q8IYB1
Gene Alias 1600021P15Rik; A430031N04; C3orf59; C87006; Mab-21 domain containing 2; Mab-21 domain containing 2a; mab-21 domain-containing protein 2; mb21d2; mb21d2a; Protein MB21D2; RGD1559643; si:ch211-170h3.5
Gene Symbols MB21D2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human C3orf59.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151963
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.