Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf75 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566690 Supplier Invitrogen™ Supplier No. PA566690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGVSLTMARER GQLVFLEGLK SAVDVVFQAQ KEPHPLQFLR EANAGNLKPL FEFVREALKP VDSGEAR Highest antigen sequence identity to the following orthologs - mouse 68%, rat 72%.

Component of the RNA polymerase II elongator complex, a multiprotein complex associated with the RNA polymerase II (Pol II) holoenzyme, and which is involved in transcriptional elongation (PubMed:22854966). The elongator complex catalyzes formation of carboxymethyluridine in the wobble base at position 34 in tRNAs (PubMed:29332244). Involved in cell migration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf75
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELP6
Gene Accession No. Q0PNE2
Gene Alias 2610001P13Rik; 2610002I17Rik; Angiotonin-transactivated protein 1; ATP1; AV159994; C3orf75; elongator acetyltransferase complex subunit 6; elongator complex protein 6; elongator complex protein 6 homolog; ELP6; Protein TMEM103; RGD1305478; Tmem103; transmembrane protein 103; UPF0405 protein C3orf75; UPF0405 protein C3orf75 homolog
Gene Symbols ELP6
Host Species Rabbit
Immunogen Recombinant Human C3orf75. Recombinant protein control fragment (Product #RP-107342).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.