Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf80 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561518 Supplier Thermo Scientific Supplier No. PA561518

Rabbit Polyclonal Antibody

Immunogen sequence: PDDDDDSPAL LRDEAAAGSQ DSLLDSGG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

C3orf80 Polyclonal antibody specifically detects C3orf80 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf80
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C3orf80
Gene Accession No. F5H4A9
Gene Alias C3orf80; chromosome 3 open reading frame 80; uncharacterized membrane protein C3orf80
Gene Symbols C3orf80
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C3orf80
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.