Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C4.4A/LYPD3 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299885R
Description
C4.4A/LYPD3 Polyclonal specifically detects C4.4A/LYPD3 in Human samples. It is validated for ELISA.Specifications
C4.4A/LYPD3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
27076 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 550 | |
C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4 | |
Produced in rabbits immunized with purified, recombinant C4.4A/LYPD3 (NP_055215.2; Met 1-His 286). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction