Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C4.4A/LYPD3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298298JF646
Description
C4.4A/LYPD3 Polyclonal antibody specifically detects C4.4A/LYPD3 in Mouse samples. It is validated for ELISASpecifications
C4.4A/LYPD3 | |
Polyclonal | |
ELISA | |
C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, Matrigel-induced gene C4 protein, MIG-C4 | |
Produced in rabbits immunized with purified, recombinant Mouse C4.4A/LYPD3 (Accession#: NP_598504.1; Met 1-His 287) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
27076 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction