Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4 gamma chain Mouse anti-Human, Clone: 2G6A7, Proteintech
SDP

Catalog No. 501736675 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6675 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6675 Supplier Proteintech Group Inc Supplier No. 662421IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody raised against 1543-1744aa of human C4-A protein recognizes C4 gamma chain.

This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene.

Specifications

Antigen C4 gamma chain
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G6A7
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C4A
Gene Accession No. P0C0L4
Gene Alias Acidic complement C4, C4, C4A, C4A2, C4A3, C4A4, C4A6, C4S, CO4, Complement C4 A, CPAMD2, RG
Gene Symbols C4A
Host Species Mouse
Immunogen C4 gamma chain Fusion Protein Ag10449
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.