Learn More
Invitrogen™ C4BPA Monoclonal Antibody (10-07)

Description
Mouse anti human C4bp monoclonal antibody, clone 051-198.6.3 (10-07), recognizes complement component 4 binding protein (C4bp). There are three variants of C4bp, composed of alpha and beta chains.
Specifications
Specifications
| Antigen | C4BPA |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 10-07 |
| Concentration | 1.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% sodium azide; pH 8.4 |
| Gene | C4BPA |
| Gene Accession No. | P04003 |
| Gene Alias | AI195242; C4b-binding protein; C4b-binding protein alpha chain; C4bp; C4bpa; complement component 4 binding protein; complement component 4 binding protein alpha; complement component 4 binding protein, alpha; proline-rich protein; PRP; RP11-164O23.4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.