Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4BPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109469 Supplier Invitrogen™ Supplier No. PA5109469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASDEHCPELP PVDNSIFVAK EVEGQILGTY VCIKGYHLVG KKTLFCNASK EWDNTTTECR LGHCPDPVLV NGEFSSSGPV NVSDKITFMC NDHYILKGSN RSQCLEDHTW APPFPICKSR DCDPPGNPVH GYFEGNNFTL GSTISYYCED RYYLVGVQEQ QCVDGEWSSA LPVCKLIQEA PKPECEKALL AFQESKNLCE AMENFMQQLK ESGMTMEELK YSLELKKAEL KAKLL.

Complement component 4-binding protein (C4BP) is a plasma glycoprotein that inhibits the classical pathway of complement activation, which is mediated through antibody targeting of foreign antigen. Structurally, C4BP is a disulfide linked, multimeric protein that is composed of seven alpha chains and one beta chain. C4BP functions as a cofactor for C3beta inactivator in the cleavage of C3beta, and accelerates the decay of C4betaC2alpha (C3 convertase) by acting as a cofactor in the cleavage of C4beta by factor I. Streptococcal strains that express Ig-binding cell surface molecules, which are members of the M protein family, can bind to overlapping C4beta binding sites in C4BP and therefore, interfere with the classical pathway of complement activation. Bacteria-bound C4BP may be an evolved mechanism that downregulates complement activation in the bacterial host microenvironment, thereby reducing the occurrences of bacterial opsonization and phagocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4BPB
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C4BPB
Gene Accession No. P20851, Q63515
Gene Alias C4b-binding protein beta chain; C4BP; C4BPB; C4bp-ps1; complement component 4 binding protein beta; complement component 4 binding protein, beta; complement component 4 binding protein, pseudogene 1
Gene Symbols C4bp-ps1, C4BPB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 17-251 of human C4BPB (NP_0010173641).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12270, 24236, 725
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.