Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563878
Description
Immunogen sequence: IMARNVSCFL VRHTPHPRRV CHIKGLNNIP ICTVNDDENA FGTLWGVGQS NYLEKNRIPF ANCSYPSSTA VQESPVRGMS PAPNGAKVP Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 52%.
The function of this protein remains unknown.
Specifications
C4orf17 | |
Polyclonal | |
Unconjugated | |
C4orf17 | |
C4orf17; chromosome 4 open reading frame 17; uncharacterized protein C4orf17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84103 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53FE4 | |
C4orf17 | |
Recombinant protein corresponding to Human C4orf17. Recombinant protein control fragment (Product #RP-100147). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction