Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4orf21 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983539
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3539 Supplier Bioss Supplier No. BS15188R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C4orf21
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZGRF1
Gene Accession No. Q86YA3
Gene Alias C4orf21; GRF-type zinc finger domain-containing protein 1; Protein ZGRF1; ZGRF1; zinc finger GRF-type containing 1; zinc finger, GRF-type containing 1
Gene Symbols ZGRF1
Host Species Rabbit
Immunogen KLH conjugated synthetic derived from human C4orf21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55345
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.