Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf22 Monoclonal Antibody (OTI1A6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526237

Catalog No. PIMA526237


Only null left
Add to Cart

Description

Description

C4orf22 Monoclonal Antibody for Western Blot, IHC (P)

C4orf22 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C4orf22
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6V702
C4orf22
Full length human recombinant protein of C4orf22 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1A6
Unconjugated
C4orf22
C4orf22; CFAP299; chromosome 4 open reading frame 22; Cilia- and flagella-associated protein 299; uncharacterized protein C4orf22
Mouse
Affinity Chromatography
RUO
255119
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.