Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144043
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144043 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144043 Supplier Invitrogen™ Supplier No. PA5144043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human MOLT-4 whole cell, human U251 whole cell, human Hela whole cell. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Probable adenosyl-L-methionine (AdoMet)-dependent tRNA (uracil-O(2)-)-methyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf23
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TRMT44
Gene Accession No. Q8IYL2
Gene Alias 2310079F23Rik; C4orf23; methyltransferase like 19; methyltransferase-like protein 19; Mettl19; probable tRNA (uracil-O(2)-)-methyltransferase; RGD1308380; TRM44; TRMT44; tRNA methyltransferase 44; tRNA methyltransferase 44 homolog; tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Symbols TRMT44
Host Species Rabbit
Immunogen E.coli-derived human TRMT44 recombinant protein (Position: M1-S757).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152992
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.