Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4orf28 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983540
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3540 Supplier Bioss Supplier No. BS15189R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PACRGL (Parkin Coregulated Like) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includebinding.

Specifications

Antigen C4orf28
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PACRGL
Gene Accession No. Q8N7B6
Gene Alias 4933428G09Rik; BOS_6501; C4orf28; LOC100343954; Pacrgl; PACRG-like protein; PARK2 coregulated like; PARK2 co-regulated like; PARK2 co-regulated-like; parkin coregulated like; RGD1311045
Gene Symbols PACRGL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C4orf28.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133015
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.