Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562457 Supplier Invitrogen™ Supplier No. PA562457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPMPLIPCLS WSTASGVFTT TDSFKMGQEF VKHFTSSADK LTNLNLVSRT LNLDISNQVV SQKPADCHNS SKTSVSATSE GLLLQDTSKM KRFNQRLSTN Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 57%.

C4orf29 (Chromosome4 open reading frame 29)is a secretedproteinin humans that is encoded by the C4orf29genewhich maps to human chromosome 4. The function of C4orf29 is uncharacterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf29
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD18
Gene Accession No. Q0P651
Gene Alias 2310068E01Rik; 3110057O12Rik; 4732404H23; ABHD18; abhydrolase domain containing 18; abhydrolase domain-containing protein 18; AI645591; Alpha/beta hydrolase domain-containing protein 18; C4orf29; hypothetical protein LOC499602; LOC499602; Protein ABHD18; RIKEN cDNA 3110057O12 gene; uncharacterized protein C4orf29; uncharacterized protein C4orf29 homolog
Gene Symbols ABHD18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C4orf29. Recombinant protein control fragment (Product #RP-100198).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80167
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.