Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525710
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525710 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525710 Supplier Invitrogen™ Supplier No. PA525710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C4orf31 Polyclonal Antibody for Flow, IHC (P), Western Blot

Promotes matrix assembly and cell adhesiveness. Promotes neuron migration, growth and survival as well as neurite outgrowth (PubMed:20969804). Promotes endothelial cell survival, vessel formation and plays an important role in the process of revascularization through NOS3-dependent mechanisms (PubMed:24706764). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf31
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NDNF
Gene Accession No. Q8TB73
Gene Alias C4orf31; fibronectin type-III domain-containing protein C4orf31; NDNF; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; NORD; Protein NDNF; UNQ2748/PRO6487
Gene Symbols NDNF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 80-109 amino acids of human C4orf31.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.