Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107004
Description
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
Promotes matrix assembly and cell adhesiveness. Promotes neuron migration, growth and survival as well as neurite outgrowth (PubMed:20969804). Promotes endothelial cell survival, vessel formation and plays an important role in the process of revascularization through NOS3-dependent mechanisms (PubMed:24706764). [UniProt].
Specifications
| C4orf31 | |
| Polyclonal | |
| Unconjugated | |
| NDNF | |
| C4orf31; fibronectin type-III domain-containing protein C4orf31; NDNF; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; NORD; Protein NDNF; UNQ2748/PRO6487 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8TB73 | |
| NDNF | |
| Synthetic peptide directed towards the C-terminal region of Human NDNF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction