Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567039 Supplier Invitrogen™ Supplier No. PA567039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GERDEDGDALA EREAAGTEWD PGASPRRRGQ RPKESEQDVE DSQN Highest antigen sequence identity to the following orthologs - mouse 39%, rat 53%.

C4orf32 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf32
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM241A
Gene Accession No. Q8N8J7
Gene Alias 2700063A17Rik; 5730508B09Rik; C4orf32; C80506; chromosome 4 open reading frame 32; FAM241A; hypothetical protein LOC691931; LOC691931; RIKEN cDNA 5730508B09 gene; uncharacterized protein C4orf32; uncharacterized protein C4orf32 homolog; Uncharacterized protein FAM241A
Gene Symbols FAM241A
Host Species Rabbit
Immunogen Recombinant Human C4orf32. Recombinant protein control fragment (Product #RP-107684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.