Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144172
Description
C4orf34 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Specifications
C4orf34 | |
Polyclonal | |
Unconjugated | |
SMIM14 | |
1110003E01Rik; 1700127H04Rik; 5430439C17Rik; C4orf34; MAd4; RGD1311122; Small integral membrane protein 14; Smim14 | |
Rabbit | |
Protein G | |
RUO | |
201895 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q96QK8 | |
SMIM14 | |
Recombinant Human Small integral membrane protein 14 protein (1-49AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction