Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C4orf36 Antibody (OTI7A7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272388FR
Description
C4orf36 Monoclonal specifically detects C4orf36 in Human samples. It is validated for Western Blot.Specifications
C4orf36 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
C4orf36 | |
Full length recombinant protein of human C4orf36 (NP_653246) produced in E.coli. | |
0.1 mL | |
132989.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI7A7 | |
Western Blot | |
chromosome 4 open reading frame 36, hypothetical protein LOC132989, MGC26744 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction