Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C4orf36 Antibody (OTI7A7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272388PE
Description
C4orf36 Monoclonal antibody specifically detects C4orf36 in Human samples. It is validated for Western BlotSpecifications
C4orf36 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
132989 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot | |
OTI7A7 | |
Western Blot | |
chromosome 4 open reading frame 36, hypothetical protein LOC132989, MGC26744 | |
Full length human recombinant protein of human C4orf36(NP_653246) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction